Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEFVKCLGHP EEFYNLVRFR VGGKRKVMPK MDQDSLSSSL KTCYKYLNQT SRSFAAVIQA LDGEMRNAVC IFYLVLRALD TLEDDMTISV EKKVPLLHNF HSFLYQPDWR FMESKEKDRQ VLEDFPTISL EFRNLAEKYQ TVIADICRRM GIGMAEFLDK HVTSEQEWDK YCHYVAGLVG IGLSRLFSAS EFEDPLVGED TERANSMGLF LQKTNIIRDY LEDQQGGREF WPQEVWSRYV KKLGDFAKPE NIDLAVQCLN ELITNALHHI PDVITYLSRL RNQSVFNFCA IPQVMAIATL AACYNNQQVF KGAVKIRKGQ AVTLMMDATN MPAVKAIIYQ YMEEIYHRIP DSDPSSSKTR QIISTIRTQN LPNCQLISRS HYSPIYLSFV MLLAALSWQY LTTLSQVTED YVQTGEH
FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Farnesyl-diphosphate farnesyltransferase; Farnesyl-diphosphate farnesyltransferase 1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase
Gene Aliases: DGPT; ERG9; FDFT1; SQS; SS
UniProt ID: (Human) P37268
Entrez Gene ID: (Human) 2222
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support