Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EIPGEELPG VCSARAFVGW YNGLPENQEL EPDLSCDTAV ILGQGNVALD VARILLTPPE HLERTDITKA ALGVLRQSRV KTVWLVGRRG PLQVAFTIKE LREMIQLPGA RPILDPVDFL GLQDKIKEVP RPRKRLTELL LRTATEKPGP AEAARQASAS RAWGLRFFRS PQQVLPSPDG RRAAGVRLAV TRLEGVDEAT RAVPTGDMED LPCGLVLSSI GYKSRPVDPS VPFDSKLGVI PNVEGRVMDV PGLYCSGWVK RGPTGVIATT MTDSFLTGQM LLQDLKAGLL PSGPRPGYAA IQALLSSRGV RPVSFSDWEK LDAEEVARGQ GTGKPREKLV DPQEMLRLLG H (142-491 aa encoded by BC063493)
This gene encodes a member of the prefoldin beta subunit family. The encoded protein is one of six subunits of prefoldin, a molecular chaperone complex that binds and stabilizes newly synthesized polypeptides, thereby allowing them to fold correctly. The complex, consisting of two alpha and four beta subunits, forms a double beta barrel assembly with six protruding coiled-coils.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenodoxin reductase; adrenodoxin-NADP(+) reductase; AR; Ferredoxin reductase; Ferredoxin--NADP(+) reductase; NADPH:adrenodoxin oxidoreductase, mitochondrial
Gene Aliases: ADXR; AR; FDXR
UniProt ID: (Human) P22570, (Rat) P56522, (Mouse) Q61578
Entrez Gene ID: (Pig) 100512919, (Human) 2232, (Rat) 79122, (Mouse) 14149
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support