Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: FLVCFGPYNV SHLVGYHQRK SPWWRSIAVV FSSLNASLDP LLFYFSSSVV RRAFGRGLQV LRNQGSSLLG RRGKDTAEGT NEDRGVGQGE GMPSSDFTTE
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fatty acid receptor 2; free fatty acid activated receptor 2; Free fatty acid receptor 2; G protein-coupled receptor 43; G-protein coupled receptor 43
Gene Aliases: FFA2; FFA2R; FFAR2; GPCR43; GPR43
UniProt ID: (Human) O15552
Entrez Gene ID: (Human) 2867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support