Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) FGF-19 is observed and less than 5% cross-reactivity with rhFGF-22 and rhFGF-23 is observed.
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-15; Fibroblast growth factor; Fibroblast growth factor 15
Gene Aliases: Fgf15; FGF19
UniProt ID: (Mouse) O35622
Entrez Gene ID: (Mouse) 14170
Molecular Function: growth factor intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support