Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FGF20 Polyclonal Antibody detects endogenous levels of FGF20 protein.
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20
Gene Aliases: FGF-20; FGF20; RHDA2
UniProt ID: (Human) Q9NP95, (Rat) Q9EST9, (Mouse) Q9ESL9
Entrez Gene ID: (Human) 26281, (Rat) 66017, (Mouse) 80857
Molecular Function:
growth factor
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support