Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PIPDSSPLLQ FGGQVRQRYL YTDDAQQTEA HLEIREDGTV GGAADQSPES LLQLKALKPG VIQILGVKTS RFLCQRPDGA LYGSLHFDPE ACSFRELLLE DGYNVYQSEA HGLPLHLPGN KSPHRDPAPR GPARFLPLPG LPPAPPEPPG ILAPQPPDVG SSDPLSMVGP SQGRSPSYAS
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196
Gene Aliases: FGF21; UNQ3115/PRO10196
UniProt ID: (Human) Q9NSA1
Entrez Gene ID: (Human) 26291
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support