Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: DCKFRERFQE NSYNTYASAI HRTEKTGREW YVALNKRGKA KRGCSPRVKP QHISTHFLPR FKQSEQPELS FTVTVPEKKK PPSPIKPKIP LSAPRKNTNS VKYRLKFRFG
Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; HBGF-5; Heparin-binding growth factor 5; Smag-82
Gene Aliases: FGF5; HBGF-5; Smag-82; TCMGLY
UniProt ID: (Human) P12034
Entrez Gene ID: (Human) 2250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support