Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SRRLIRTYQL YSRTSGKHVQ VLANKRINAM AEDGDPFAKL IVETDTFGSR VRVRGAETGL YICMNKKGK
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system. Plays a role in neurite outgrowth in hippocampal cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIGF; androgen-induced; Androgen-induced growth factor; FGF; FGF-8; Fibroblast growth factor; Fibroblast growth factor 8; fibroblast growth factor 8 (androgen-induced); HBGF-8; Heparin-binding growth factor 8; MGC149376
Gene Aliases: AIGF; FGF-8; FGF8; HBGF-8; HH6; KAL6
UniProt ID: (Human) P55075
Entrez Gene ID: (Human) 2253
Molecular Function:
growth factor
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support