Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Acidic and basic fibroblast growth factors (FGFs) are members of a family of multifunctional polypeptide growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuro-ectodermal origin. Like other growth factors, FGFs act by binding and activating specific cell surface receptors. A total of six members of the FGF receptor family have been identified and cloned. These include the Flg receptor or FGFR-1, the BEK receptor or FGFR-2, FGFR-3, FGFR-4, FGFR-5 and FGFR-6. These receptors usually contain an extracellular ligand-binding region containing three immunoglobulin-like domains, a transmembrane domain and a cytoplasmic tyrosine kinase domain. However, FGFR-5 lacks the cytoplasmic kinase domain. FGFR-5 is expressed in a broad range of tissues, including kidney, brain and lung, but it is preferentially expressed in pancreas, where it may play a role in the regulation of some pancreatic function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-L-iduronidase; FGF homologous factor receptor; FGF receptor-like protein 1; FGFR-5; FGFR-like protein; Fibroblast growth factor receptor 5; Fibroblast growth factor receptor-like 1; fibroblast growth factor receptor-like protein
Gene Aliases: FGFR5; FGFR5beta; FGFR5gamma; FGFRL1; FHFR; UNQ480/PRO943
UniProt ID: (Human) Q8N441, (Mouse) Q91V87, (Rat) Q7TQM3
Entrez Gene ID: (Human) 53834, (Mouse) 116701, (Rat) 360903
Molecular Function:
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support