Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EYESSFSNEE AQKAQALRLA SHLNLAMCHL KLQAFSAAIE SCNKALELDS NNEKGLFRRG EAHLAVNDFE LARADFQKVL QLYPNNKAAK TQLAVCQQRI RRQLAREKKL
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 51 kDa FK506-binding protein; 52 kDa FK506-binding protein; 52 kDa FKBP; 59 kDa immunophilin; FK506 binding protein 4, 59kDa; FK506 binding protein 52; FK506-binding protein 4; FK506-binding protein 4 (59kD); FKBP-4; FKBP51; FKBP52 protein; FKBP59; HBI; HSP binding immunophilin; HSP-binding immunophilin; Immunophilin FKBP52; p59; Peptidyl-prolyl cis-trans isomerase FKBP4; peptidylprolyl cis-trans isomerase; PPIase FKBP4; Rotamase; T-cell FK506-binding protein, 59kD
Gene Aliases: 59kDa; AL022792; AW208983; FKBP-4; FKBP-52; FKBP4; FKBP51; FKBP52; FKBP59; FKPB52; HBI; Hsp56; p52; p59; PPIase
UniProt ID: (Human) Q02790, (Mouse) P30416, (Rat) Q9QVC8
Entrez Gene ID: (Human) 2288, (Mouse) 14228, (Rat) 260321
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support