Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGQPPAEEA EQPGALAREF LAAMEPEPAP APAPEEWLDI LGNGLLRKKT LVPGPPGSSR PVKGQVVTVH LQTSLENGTR VQEEPELVFT LGDCDVIQAL DLSVPLMDVG ETAMVTADSK YCYGPQGRSP YIPPHAALCL EVTLKTAVDG PDLEMLTGQE RVALANRKRE CGNAHYQRAD FVLAANSYDL AIKAITSSAK VDMTFEEEAQ LLQLKVKCLN NLAASQLKLD HYRAALRSCS LVLEHQPDNI KALFRKGKVL AQQGEYSEAI PILRAALKLE PSNKTIHAEL SKLVKKHAAQ RSTETALYRK MLGNPSRLPA KCPGKGAWSI PWKWLFGATA VALGGVALSV VIAARN (1-355 aa encoded by B C009966)
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 38 kDa FK506-binding protein; 38 kDa FKBP; FK506 binding protein 8, 38kDa; FK506-binding protein 38; FK506-binding protein 8; FK506-binding protein 8 (38kD); FKBP-38; FKBP-8; FKBPR38; hFKBP38; mFKBP38; Peptidyl-prolyl cis-trans isomerase FKBP8; PPIase FKBP8; Rotamase
Gene Aliases: 38kDa; FKBP-38; FKBP-8; FKBP38; FKBP8; FKBPr38; Sam11
UniProt ID: (Human) Q14318, (Mouse) O35465
Entrez Gene ID: (Human) 23770, (Mouse) 14232
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support