Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ETPPVNTIG EKDTSQPQQE WEKNLRENLD SVIQIRQQPR DPPTETLELE VSPDPASQIL EHTQGAEKLV AELEGDSHKS HGSTSQMPEA LQASDLWYCP DGSFVKKIVI RGHGLDKPKL GSCCRVLALG FPFGSGPPEG WTELTMGVGP WREETWGELI EKCLESMCQG EEAELQLPGH SGPPVRLTLA SFTQGRDSWE LETSEKEALA REERARGTEL FRAGNPEGAA RCYGRAL (2-237 aa encoded by BC004168)
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FK506-binding protein like; FK506-binding protein-like; immunophilin-like protein NG7; peptidyl-prolyl cis-trans isomerase; WAF-1/CIP1 stabilizing protein 39; WISp39
Gene Aliases: DIR1; FKBP4; FKBPL; NG7; Ppiase-X; WISP39
UniProt ID: (Human) Q9UIM3, (Mouse) O35450, (Rat) Q6MG81
Entrez Gene ID: (Human) 63943, (Mouse) 56299, (Rat) 406168
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support