Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Muscle
Cellular location: Cytoplasmic.
This gene encodes a protein which is targeted to the medial Golgi apparatus and is necessary for posttranslational modification of dystroglycan. Mutations in this gene have been associated with congenital muscular dystrophy, mental retardation, and cerebellar cysts. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq, Oct 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fukutin-related protein; Ribitol 5-phosphate transferase FKRP; Ribitol-5-phosphate transferase
Gene Aliases: A830029B19Rik; AI842067; AI847300; FKRP; LGMD1I; LGMD2I; MDC1C; MDDGA5; MDDGB5; MDDGC5
UniProt ID: (Human) Q9H9S5, (Mouse) Q8CG64
Entrez Gene ID: (Human) 79147, (Mouse) 243853, (Rat) 308390
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support