Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNAIVALCHF CELHGPRTLF CTEVLHAPLP QGDGNEDSPG QGEQAEEEEG GIQMNSRMRA HSPAEGASVE SSSPGPKKSD MCEGCRSLAA GHPGYISHDK ETSIKYVSHQ HPSHPQLFSI VRQACVRSLS CEVCPGREGP IFFGDEQHGF VFSHTFFIKD SLARGFQRWY SIITIMMDRI YLINSWPFLL GKVRGIIDEL QGKALKVFEA EQFGCPQRAQ RMNTAFTPFL HQRNGNAARS LTSLTSDDNL WACLHTSFAW LLKACGSRLT EKLLEGAPTE DTLVQMEKLA GEAGVLLPGP WPGWPWGGTS CLLSWQESLR EGNAALNQPR TSLREAHPPI SV
This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein; Folliculin
Gene Aliases: BHD; FLCL; FLCN
UniProt ID: (Human) Q8NFG4
Entrez Gene ID: (Human) 201163
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support