Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MWLDHRAVSQ VNRINETKHS VLQYVGGVMS VEMQAPKLLW LKENLREICW DKAGHFFDLP DFLSWKATGV TARSLCSLVC KWTYSAEKGW DDSFWKMIG
FGGY gene ontology annotations related to this gene include neuron cellular homeostasis; pentose metabolic process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-ribulokinase FGGY; FGGY carbohydrate kinase domain-containing protein
Gene Aliases: FGGY
UniProt ID: (Human) Q96C11
Entrez Gene ID: (Human) 55277
Molecular Function: carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support