Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRLKLFSILS TVLLRATDTI NSQGQFPSYL ETVTKDILAP NLQWHAGRTA AAIRTAAVSC LWALTSSEVL SAEQIRDVQE TLMPQVLTTL EEDSKMTRLI SCRIINTFLK TSGGMTDPEK LIKIYPELLK RLDDVSNDVR MAAASTLVTW LQCVKGANAK SYYQSSVQYL YRELLVHLDD PERAIQDAIL DVPGSPSPSA MIGSFLRPPH PWRTVSQLNL FSS
The protein encoded by this gene is essential for the preassembly or stability of axonemal dynein arms, and is found only in organisms with motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia-18, a disorder characterized by abnormalities of motile cilia. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynein (axonemal) assembly factor 5; Dynein axonemal assembly factor 5; dynein, axonemal, assembly factor 5; HEAT repeat containing 2; HEAT repeat-containing protein 2
Gene Aliases: CILD18; DNAAF5; HEATR2
UniProt ID: (Human) Q86Y56
Entrez Gene ID: (Human) 54919
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support