Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNIHRALADI LRQQGPIPIA HCERETISAI DTSPKENTPV RSSSKNHYTP VRTAKQTPEK PRMNNILTSA TEPYDLSFSR SFQNLAHLPP SYESAVKTNP SKYSSLKRLT DKEADEYYMR RRHLPDLAAR GTLPLNVIQM SQQKPLPRER PRRPIRAMSQ DRVLSPDRGL PDEFSMPYDR ILSDEQLLST ERLHSQDPLL SPERTAFPEQ SLSRAISHTD VFVSTPVLDR YRMSKMHSHP SASNNSYATL GQSQTAAKRH AFASRRHNTV EQLHYIPGHH TCYTASKTEV TV
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein shisa-6; shisa homolog 6
Gene Aliases: SHISA6
UniProt ID: (Human) Q6ZSJ9
Entrez Gene ID: (Human) 388336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support