Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABP-280; actin binding protein 280; Actin-binding protein 280; alpha filamin; Alpha-filamin; Endothelial actin-binding protein; filamin 1; filamin A, alpha; Filamin-1; Filamin-A; FilaminA; FLN-A; Non-muscle filamin
Gene Aliases: ABP-280; ABPX; CSBS; CVD1; FLN; FLN-A; FLN1; FLNA; FMD; MNS; NHBP; OPD; OPD1; OPD2; XLVD; XMVD
UniProt ID: (Human) E9KL45
Entrez Gene ID: (Human) 2316
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support