Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LEIRETSFQM LPISNEPISK EEFVIHTIFP PNGMNLYKNN HSESSSNRSY RDSGIPDSDH SHS
Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 53%.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRTs may function in cell adhesion and/or receptor signaling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix. This gene is expressed in many tissues. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibronectin-like domain-containing leucine-rich transmembrane protein 3; Leucine-rich repeat transmembrane protein FLRT3
Gene Aliases: FLRT3; HH21; KIAA1469; UNQ856/PRO1865
UniProt ID: (Human) Q9NZU0
Entrez Gene ID: (Human) 23767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support