Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTVLAPAWSP TTYLLLLLLL SSGLSGTQDC SFQHSPISSD FAVKIRELSD YLLQDYPVTV ASNLQDEELC GGLWRLVLAQ RWMERLKTVA GSKMQGLLER VNTEIHFVTK CAFQPPPSCL RFVQTNISRL LQETSEQLVA LKPWITRQNF SRCLELQCQP DSSTLPPPWS PRPLEATAPT APQPPLLLLL LLPVGLLLLA AAWCLHWQRT RRRTPRPGEQ VPPVPSPQDL LLVEH
FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Flt 3 ligand; Flt3 L; Flt3 ligand; Fms-related tyrosine kinase 3 ligand; H-FLT-3-L; M-FLT-3-L; SL cytokine
Gene Aliases: FL; FLT3L; FLT3LG
UniProt ID: (Human) P49771
Entrez Gene ID: (Human) 2323
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support