Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: NIDMPKTEPK GADPESPRRE EMGCNADQES QSGPGVPQTQ GGEVKPKSPE TALEAFKALF IRPPRKGTTA DTSELEALKR KMRHEKESLR AVFERSNSKP ADGPSDSKSP DHSLTEQDDR TPGRLQAVWP PPKTKDTEEK VGLKYT; Positive Samples: 293T, A-549, U-87MG, LO2, Mouse testis; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Cytoplasmic side, Nucleus, Peripheral membrane protein, adherens junction
Plays a role in the formation of adherens junction and the polymerization of linear actin cables.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: formin (limb deformity); Formin-1; Limb deformity protein; Limb deformity protein homolog
Gene Aliases: FMN; FMN1; LD
UniProt ID: (Human) Q68DA7, (Mouse) Q05860
Entrez Gene ID: (Human) 342184, (Mouse) 14260
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support