Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DLHFLDKAGS VSLDSVLADV RSLQRGLELT QREFVRQDDC MVLKEFLRAN SPTMDKLLAD SKTAQEAFES VVEYFGENPK TTSPGLFFSL FSRFIKAYKK AEQEVEQWKK EAAAQEAGAD TPGKGEPPAP KSPPKARRPQ MDLISELKRR QQKEPLIYES DRDGAIEDII T; Positive Samples: LO2, A-549, Mouse brain; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Cytoplasmic vesicle, Lipid-anchor, bleb, cell cortex, phagosome
The Formin-like protein 1 (FRL1) gene encodes a formin-related protein which has been implicated in morphogenesis, cyokinesis, and cell polarity. Formins are a conserved class of proteins expressed in all eukaryotes and have one DAD (diaphanous autoregulatory domain), one FH2 (formin homology 2) domain and one GBD/FH3 (Rho GTPase-binding / formin homology 3) domain. FRL1 is located in the cytoplasm and is highly expressed in the spleen, lymph node and bone marrow cells. FRL1 possibly has a role in the control of cell motility, survival of macrophages and cytoskeletal organization.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLL-associated antigen KW-13; formin-like 1; Formin-like protein 1; formin-related gene in leukocytes; Formin-related protein; Leukocyte formin; lymphocyte specific formin related protein
Gene Aliases: 8030453N10Rik; AI553564; C17orf1; C17orf1B; FHOD4; FMNL; FMNL1; Fnrl; Frl; FRL1; Frls; KW-13
UniProt ID: (Human) O95466, (Mouse) Q9JL26
Entrez Gene ID: (Human) 752, (Mouse) 57778
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support