Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FNDC3B may be a positive regulator of adipogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Factor for adipocyte differentiation 104; fibronectin domain-containing protein 3b; Fibronectin type III domain-containing protein 3B; HCV NS5A-binding protein 37
Gene Aliases: 1600019O04Rik; AW550168; FAD104; FNDC3B; Kiaa4164; mKIAA4164; NS5ABP37; PRO4979; RGD1311673; UNQ2421/PRO4979/PRO34274; YVTM2421
UniProt ID: (Human) Q53EP0, (Mouse) Q6NWW9
Entrez Gene ID: (Human) 64778, (Rat) 294925, (Mouse) 72007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support