Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: EDVGALDCGS PASSTPYFTG LELPGELKLD APYNFNHPFS INNLMSEQTP APPKLDVGFG GYGAEGGEPG VYYQGLYSRS LLNAS
FOXA3 encodes a member of the forkhead class of DNA-binding proteins. These hepatocyte nuclear factors are transcriptional activators for liver-specific transcripts such as albumin and transthyretin, and they also interact with chromatin. Similar family members in mice have roles in the regulation of metabolism and in the differentiation of the pancreas and liver. The crystal structure of a similar protein in rat has been resolved.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fork head-related protein FKH H3; Forkhead box protein A3; hepatocyte nuclear factor 3, gamma; Hepatocyte nuclear factor 3-gamma; HNF-3-gamma; HNF-3G; TCF-3G; Transcription factor 3G
Gene Aliases: FKHH3; FOXA3; HNF3G; TCF3G
UniProt ID: (Human) P55318
Entrez Gene ID: (Human) 3171
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support