Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transcription factor required for formation of positional identity in the developing retina, regionalization of the optic chiasm and morphogenesis of the kidney. Can neutralize ectodermal cells directly. Binds to the consensus sequence 5'-A[AT]T[AG]TTTGTTT-3' and acts as a transcriptional repressor. Also acts as a transcriptional activator. Promotes development of neural crest cells from neural tube progenitors. Restricts neural progenitor cells to the neural crest lineage while suppressing interneuron differentiation. Required for maintenance of pluripotent cells in the pre-implantation and peri-implantation stages of embryogenesis. Probable transcription factor involved in embryogenesis and somatogenesis. FOXD1 is involved in regulating inflammation as well as kidney and retinal development. FOXD1 regulates the activity of NFAT and NFkB. Deficiency of FOXD1 results in multiorgan systemic inflammation, exaggerated Th cell-derived cytokine production, and T cell proliferation in autogolgous MLRs. In kidneys, FOXD1 controls the production of signals required for the normal transition of induced mesenchyme into tubular epithelium and full growth and branching of the collecting system. Deletion of FOXD1 results in renal abnormalities. FOXD2 acts as a modulator of T cell activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BF-2; Brain factor 2; Forkhead box protein D1; FORKHEAD BOX PROTEIN D1 (FORKHEAD-RELATED PROTEIN FKHL8) (FORKHEAD-RELATED TRANSCRIPTION FACTOR 4) (FREAC-4); Forkhead box protein D2; Forkhead box protein D3; Forkhead box protein D4; forkhead homolog 2; forkhead, drosophila, homolog-like 17; Forkhead, drosophila, homolog-like 8; forkhead, Drosophila, homolog-like 9; forkhead-like 17; forkhead-like 8; forkhead-related activator 4; forkhead-related activator 5; forkhead-related activator 9; Forkhead-related protein FKHL17; Forkhead-related protein FKHL8; Forkhead-related protein FKHL9; Forkhead-related transcription factor 4; Forkhead-related transcription factor 5; Forkhead-related transcription factor 9; FREAC-4; FREAC-5; FREAC-9; Hepatocyte nuclear factor 3 forkhead homolog 2; HFH-2; HFH-BF-2; HNF-3/forkhead homolog 2; HNF-3/forkhead homolog, brain factor 2; HNF3/FH transcription factor genesis; Mesoderm/mesenchyme forkhead 2; MF-2; Myeloid factor-alpha; Transcription factor FKH-2
Gene Aliases: AI385632; AI426778; AIS1; BF-2; CWH3; Fkh2; FKHL17; FKHL8; FKHL9; FOXD1; FOXD2; FOXD3; FOXD4; FOXD4A; FREAC-4; FREAC-5; FREAC-9; FREAC4; FREAC5; FREAC9; Genesis; Hfh10; HFH2; Hfhbf2; Mf2; VAMAS2
UniProt ID: (Human) Q16676, (Human) O60548, (Human) Q9UJU5, (Human) Q12950, (Mouse) Q61345, (Mouse) O35392, (Mouse) Q61060, (Mouse) Q60688
Entrez Gene ID: (Human) 2297, (Human) 2306, (Human) 27022, (Human) 2298, (Mouse) 15229, (Mouse) 17301, (Mouse) 15221, (Mouse) 14237
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support