Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to FOXH1.
FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fast-1; Fast-2; FAST1 transcription factor; Forkhead activin signal transducer 1; Forkhead activin signal transducer 2; forkhead activin signal transducer-1; Forkhead box protein H1; hFAST-1; TGF-beta/activin signal transducer
Gene Aliases: FAST-1; fast-2; FAST1; FAST2; FOXH1
UniProt ID: (Human) O75593, (Mouse) O88621
Entrez Gene ID: (Human) 8928, (Rat) 300054, (Mouse) 14106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support