Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAESWLRLSG AGPAEEAGPE GGLEEPDALD DSLTSLQWLQ EFSILNAKAP ALPPGGTDPH GYHQVPGSAA PGSPLAADPA CLGQPHTPGK PTSSCTSRSA PPGLQAPPPD DVDYATNPHV KPPYSYATLI CMAMQASKAT KITLSAIYKW ITDNFCYFRH ADPTWQNSIR HNLSLNKCFI KVPREKDEPG KGGFWRIDPQ YAERLLSGAF KKRRLPPVHI HPAFARQAAQ EPSAVPRAGP LTVNTEAQQL LREFEEATGE AGWGAGEGRL GHKRKQPLPK RVAKVPRPPS TLLPTPEEQG ELEPLKGNFD WEAIFDAGTL GGELGALEAL ELSPPLSPAS HVDVDLTIHG RHIDCPATWG PSVEQAADSL DFDETFLATS FLQHPWDESG SGCLPPEPLF EAGDATLASD LQDWASVGAF L
Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fork head homologue 4; Forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4; MGC35202
Gene Aliases: FKHL13; FOXJ1; HFH-4; HFH4
UniProt ID: (Human) Q92949
Entrez Gene ID: (Human) 2302
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support