Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSHLFDPRLP ALAASPMLYL YGPERPGLPL AFAPAAALAA SGRAETPQKP PYSYIALIAM AIQDAPEQRV TLNGIYQFIM DRFPFYHDNR QGWQNSIRHN LSLNDCFVKV PREKGRPGKG SYWTLDPRCL DMFENGNYRR RKRKPKPGPG APEAKRPRAE THQRSAEAQP EAGSGAGGSG PAISRLQAAP AGPSPLLDGP SPPAPLHWPG TASPNEDAGD AAQSAAAVAV GQAARTGDGP GSPLRPASRS SPKSSDKSKS FSIDSILAGK QGQKPPSGDE LLGGAKPGPG GRLGASLLAA SSSLRPPFNA SLMLDPHVQG GFYQLGIPFL SYFPLQVPDT VLHFQ
The forkhead domain is a 100-amino acid monomeric DNA binding motif originally identified as a region of homology between the Drosophila forkhead protein and rat HNF3. Pierrou et al. (1994) identified 7 human genes containing forkhead domains and designated them forkhead related activators (FREAC) 1 through 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FKH11; FKHL10; Forkhead box protein L1; Forkhead related transcription factor; forkhead-like 11; Forkhead-related protein FKHL11; Forkhead-related transcription factor 7; FREAC-7; FREAC6; JFJ-3
Gene Aliases: FKH6; FKHL11; FOXL1; FREAC7
UniProt ID: (Human) Q12952
Entrez Gene ID: (Human) 2300
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support