Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to the C-term of FOXL2.
FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species. Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation in mammals and in ovarian somatic cell differentiation and in further follicle development and/or maintenance. Immunohistochemical data shows that FOXL2 is a nuclear protein specifically expressed in eyelids and in fetal and adult ovarian follicular cells. It does not undergo any major posttranslational modifications. FOXL2 is also found to be associated with blepharo-phimosis/ptosis/epicanthus inverses syndrome (BPES). There are two forms of BPES. Type I BPES shows eyelid abnormal-ities that are associated with ovarian failure. In type II BPES, only eyelid defects are found. Studies have shown that FOXL2 is expressed in the mesenchyme of developing mouse eyelids and in adult ovarian follicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Blepharophimosis; epicanthus inversus; forkhead box L2; Forkhead box protein L2; forkhead transcription factor FOXL2; LUSTR2; MGC14393; P-Frk; Pituitary forkhead factor; putative transcription factor foxl2
Gene Aliases: AU045128; BPES; BPES1; FOXL2; PFRK; PINTO; POF3
UniProt ID: (Human) P58012, (Mouse) O88470
Entrez Gene ID: (Human) 668, (Mouse) 26927, (Rat) 367152
Molecular Function:
winged helix/forkhead transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support