Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMQESATET ISNSSMNQNG MSTLSSQLDA GSRDGRSSGD TSSEVSTVEL LHLQQQQ (1-56 aa encoded by B C018016)
FOXP2 is a member of the forkhead/winged-helix (FOX) family of transcription factors. It is expressed in fetal and adult brain as well as in several other organs such as the lung and gut. The protein product contains a FOX DNA-binding domain and a large polyglutamine tract and is an evolutionarily conserved transcription factor, which may bind directly to approximately 300 to 400 gene promoters in the human genome to regulate the expression of a variety of genes. This gene is required for proper development of speech and language regions of the brain during embryogenesis, and may be involved in a variety of biological pathways and cascades that may ultimately influence language development. Mutations in this gene cause speech-language disorder 1 (SPCH1), also known as autosomal dominant speech and language disorder with orofacial dyspraxia. Multiple alternative transcripts encoding different isoforms have been identified in this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAG repeat protein 44; DKFZp686H1726; Forkhead box protein P2; forkhead/winged-helix transcription factor; OTTHUMP00000196932; trinucleotide repeat containing 10; Trinucleotide repeat-containing gene 10 protein
Gene Aliases: 2810043D05Rik; AI449000; CAG-16; CAGH44; D0Kist7; FOXP2; RGD1559697; SPCH1; TNRC10
UniProt ID: (Human) O15409, (Mouse) P58463, (Rat) P0CF24
Entrez Gene ID: (Human) 93986, (Mouse) 114142, (Rat) 500037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support