Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ40908; FLJ44184; fork head-related protein like A; Fork head-related protein-like A; Forkhead box protein P4; foxp4; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4
Gene Aliases: 1200010K03Rik; 2310007G05Rik; FKHLA; FOXP4; hFKHLA; mFKHLA
UniProt ID: (Human) Q8IVH2, (Mouse) Q9DBY0
Entrez Gene ID: (Human) 116113, (Mouse) 74123
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support