Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METNFSTPLN EYEEVSYESA GYTVLRILPL VVLGVTFVLG VLGNGLVIWV AGFRMTRTVT TICYLNLALA DFSFTATLPF LIVSMAMGEK WPFGWFLCKL IHIVVDINLF GSVFLIGFIA LDRCICVLHP VWAQNHRTVS LAMKVIVGPW ILALVLTLPV FLFLTTVTIP NGDTYCTFNF ASWGGTPEER LKVAITMLTA RGIIRFVIGF SLPMSIVAIC YGLIAAKIHK KGMIKSSRPL RVLTAVVASF FICWFPFQLV ALLGTVWLKE MLFYGKYKII DILVNPTSSL AFFNSCLNPM LYVFVGQDFR ERLIHSLPTS LERALSEDSA PTNDTAANSA SPPAETELQA M
Formyl peptide receptor 2 (FPR2) is a low affinity receptor for N-formyl-methionyl peptides. Binding of FMLP to FPR2 causes activation of neutrophils. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FMLP-R-I; FMLP-related receptor I; fMLPR1; Formyl peptide receptor-like 1; FPR-like 1; Fpr-rs1; Fprl-1; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); Lipoxin receptor; LXA4 receptor; N-formyl peptide receptor 2; RFP
Gene Aliases: ALXR; FMLP-R-II; FMLPX; FPR2; FPR2A; FPRH1; FPRH2; FPRL1; HM63; LXA4R
UniProt ID: (Human) P25090
Entrez Gene ID: (Human) 2358
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support