Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 2.7.1.112; EC 2.7.10.2; FYN-related kinase; fyn-related Src family tyrosine kinase; Nuclear tyrosine protein kinase RAK; Protein-tyrosine kinase 5; PTK5 protein tyrosine kinase 5; Tyrosine-protein kinase FRK
Gene Aliases: FRK; GTK; PTK5; RAK
UniProt ID: (Human) P42685
Entrez Gene ID: (Human) 2444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support