Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EEWADLFQA AGAKYVVLTT KHHEGFTNWP SPVSWNWNSK DVGPHRDLVG ELGTALRKRN IRYGLYHSLL EWFHPLYLLD KKNGFKTQHF VSAKTMPELY DLVNSYKPDL IWSDGEWECP DTYWNSTNFL SWLYNDSPVK DEVVVNDRWG QNCSCHHGGY YNCEDKFKPQ SLPDHKWEMC TSIDKFSWGY RRDMALSDVT EESEIISELV QTVSLGGNYL LNIGPTKDGL IVPIFQERLL AVGKWLSING EAIYASKPWR VQWEKNTTSV WYTSKGSAVY AIFLHWPENG VLNLESPITT STTKITMLGI QGDLKWSTDP DKGLFISLPQ LPPSAVPAEF AWTIKLTGVK (113-461 aa encoded by BC017338)
Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids (Occhiodoro et al., 1989 [PubMed 2803312]). At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis (MIM 230000), and that in plasma, FUCA2 (MIM 136820).[supplied by OMIM].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A L fucosidase I; A L fucoside fucohydrolase 1; Alpha-L-fucosidase 1; Alpha-L-fucosidase I; Alpha-L-fucoside fucohydrolase 1; EC 3.2.1; EC 3.2.1.51; Fucosidase alpha-L-1 tissue; Fucosidase, alpha-L-1, tissue; Tissue alpha-L-fucosidase; Tissue α L fucosidase; α L fucosidase 1; α L fucosidase I; α L fucoside fucohydrolase 1
Gene Aliases: 0610006A03Rik; 9530055J05Rik; Afuc; FUCA; FUCA1; Nbla10230
UniProt ID: (Human) P04066, (Rat) P17164, (Mouse) Q99LJ1
Entrez Gene ID: (Human) 2517, (Rat) 24375, (Mouse) 71665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support