Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGAKYIVLT SKHHEGFTLW GSEYSWNWNA IDEGPKRDIV KELEVAIRNR TDLRFGLYYS LFEWFHPLFL EDESSSFHKR QFPVSKTLPE LYELVNNYQP EVLWSDGDGG APDQYWNSTG FLAWLYNESP VRGTVVTNDR WGAGSICKHG GFYTCSDRYN PGHLLPHKWE NCMTIDKLSW GYRREAGISD YLTIEELVKQ LVETVSCGGN LLMNIGPTLD GTISVVFEER LRQVGSWLKV NGEAIYETYT WRSQNDTVTP DVWYTSKPKE KLVYAIFLKW PTSGQLFLGH PKAILGATEV KLLGHGQPLN WISLEQNGIM VELPQLTIHQ MPCKWGWALA LTNVI (124-467 aa encoded by BC003060)
FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-L-fucosidase 2; Alpha-L-fucoside fucohydrolase 2; MGC1314; Plasma alpha-L-fucosidase
Gene Aliases: 0610025O11Rik; 5530401P20Rik; dJ20N2.5; FUCA2; PSEC0151; UNQ227/PRO260
UniProt ID: (Human) Q9BTY2, (Mouse) Q99KR8, (Rat) Q6AYS4
Entrez Gene ID: (Human) 2519, (Mouse) 66848, (Rat) 292485
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support