Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRPQELPRLA FPLLLLLLLL LPPPPCPAHS ATRFDPTWES LDARQLPAWF DQAKFGIFIH WGVFSVPSFG SEWFWWYWQK EKIPKYVEFM KDNYPPSFKY EDFGPLFTAK FFNANQWADI FQASGAKYIV LTSKHHEGFT LWGSEYSWNW NAIDEGPKRD IVKELEVAIR NRTDLRFGLY YSLFEWFHPL FLEDESSSFH KRQFPVSKTL PELYELVNNY QPEVLWSDGD GGAPDQYWNS TGFLAWLYNE SPVRGTVVTN DRWGAGSICK HGGFYTCSDR YNPGHLLPHK WENCMTIDKL SWGYRREAGI SDYLTIEELV KQLVETVSCG GNLLMNIGPT LDGTISVVFE ERLRQVGSWL KVNGEAIYET YTWRSQNDTV TPDVWYTSKP KEKLVYAIFL KWPTSGQLFL GHPKAILGAT EVKLLGHGQP LNWISLEQNG IMVELPQLTI HQMPCKWGWA LALTNVI
FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-L-fucosidase 2; Alpha-L-fucoside fucohydrolase 2; MGC1314; Plasma alpha-L-fucosidase
Gene Aliases: dJ20N2.5; FUCA2; PSEC0151; UNQ227/PRO260
UniProt ID: (Human) Q9BTY2
Entrez Gene ID: (Human) 2519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support