Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWLRSHRQLC LAFLLVCVLS VIFFLHIHQD SFPHGLGLSI LCPDRRLVTP PVAIFCLPGT AMGPNASSSC PQHPASLSGT WTVYPNGRFG NQMGQYATLL ALAQLNGRRA FILPAMHAAL APVFRITLPV LAPEVDSRTP WRELQLHDWM SEEYADLRDP FLKLSGFPCS WTFFHHLREQ IRREFTLHDH LREEAQSVLG QLRLGRTGDR PRTFVGVHVR RGDYLQVMPQ RWKGVVGDSA YLRQAMDWFR ARHEAPVFVV TSNGMEWCKE NIDTSQGDVT FAGDGQEATP WKDFALLTQC NHTIMTIGTF GFWAAYLAGG DTVYLANFTL PDSEFLKIFK PEAAFLPEWV GINADLSPLW TLAKP
FUT1 is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2-alpha-L-fucosyltransferase; alpha (1,2) fucosyltransferase; alpha(1,2) fucosyltransferase 1; Alpha(1,2)FT 1; Blood group H alpha 2-fucosyltransferase; Fucosyltransferase 1; fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase); fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group); Galactoside alpha-(1,2)-fucosyltransferase 1; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 1; Type 1 galactoside alpha-(1,2)-fucosyltransferase FUT1; Type 2 galactoside alpha-(1,2)-fucosyltransferase FUT1
Gene Aliases: FUT1; H; HH; HSC
UniProt ID: (Human) P19526
Entrez Gene ID: (Human) 2523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support