Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALLHEESPL NNFLLSHGPG IRLFNLTSTF SRHSDYPLSL QWLPGTAYLR RPVPPPMERA EWRRRGYAPL LYLQSHCDVP ADRDRYVREL MRHIPVDSYG KCLQNRELPT ARLQDTATAT TEDPELLAFL SRYKFHLALE NAICNDYMTE KLWRPMHLGA VPVYRGSPSV RDWMPNNHSV ILIDDFESPQ KLAEFIDFLD KNDEEYMKYL AYKQPGGITN QFLLDSLKHR EWGVNDPLLP NYLNGFECFV CDYELARLDA EKAHAASPGD SPVFEPHIAQ PSHMDCPVPT PGFGNVEEIP ENDSWKEMWL QDYWQGLDQG EALTAMIHNN ETEQTKFWDY LHEIFMKRQH L (143-492 aa encoded by BC036037)
Probable fucosyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha (1,3) fucosyltransferase; Alpha-(1,3)-fucosyltransferase 11; alpha3-fucosyltransferase 11; EC 2.4.1; Fuc-TXI; Fucosyltransferase 11; fucosyltransferase 11 (alpha (1,3) fucosyltransferase); Fucosyltransferase XI; fucT-XI; Galactoside 3-L-fucosyltransferase 11; MGC119338; MGC119339; MGC33202
Gene Aliases: 3110009G03Rik; AI317183; FUT11
UniProt ID: (Human) Q495W5, (Mouse) Q8BHC9, (Rat) Q68FV3
Entrez Gene ID: (Human) 170384, (Mouse) 73068, (Rat) 286971
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support