Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; FXR1P; hFXR1p; mFxr1p; RNA-binding protein FXR1
Gene Aliases: 1110050J02Rik; 9530073J07Rik; AI851072; FXR1; Fxr1h; FXR1P
UniProt ID: (Human) P51114, (Mouse) Q61584
Entrez Gene ID: (Human) 8087, (Mouse) 14359
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support