Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -4, -7, or -8 is observed. By flow cytometry and immunohistochemistry, this antibody also detects mouse Frizzled-3 on differentiated embryoid bodies derived from the mouse embryonic cell line, D3. This antibody also detects Frizzled-3 on human HEK293 cells by immunocytochemistry and flow cytometry.
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. The function of this protein is unknown, although it may play a role in mammalian hair follicle development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frizzled 3, seven transmembrane spanning receptor; frizzled family receptor 3; frizzled homolog 3; Frizzled-3; Fz-3; hFz3; mFz3
Gene Aliases: AU020229; D930050A07Rik; Fz-3; Fz3; FZD3
UniProt ID: (Human) Q9NPG1, (Mouse) Q61086
Entrez Gene ID: (Human) 7976, (Mouse) 14365
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support