Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GKTVESWRR FTSRCCCRPR RGHKSGGAMA AGDYPEASAA LTGRTGPPGP AATYHKQVSL SHV (524-585 aa encoded by BC114346)
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled family receptor 5; Frizzled-5; Fz-5; Fz5; Fzd-5; FzE5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Aliases: C2orf31; FZD5; HFZ5
UniProt ID: (Human) Q13467
Entrez Gene ID: (Human) 7855
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support