Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TSQVTRLCD LSVEGDSVTS VGWSERGNLV AVGTHKGFVQ IWDAAAGKKL SMLEGHTARV GALAWNAEQL SSGSRDRMIL QRDIRTPPLQ SERRLQGHRQ EVCGLKWSTD HQLLASGGND NKLLVWNHSS LSPVQQYTEH LAAVKAIAWS PHQHGLLASG GGTADRCIRF WNTLTGQPLQ CIDTGSQVCN LAWSKHANEL VSTHGYSQNQ ILVWKYPSLT QVAKLTGHSY RVLYLAMSPD GEAIVTGAGD ETLRFWNVFS KTRSTKESVS VLNLFTRIR (216-493 aa encoded by BC013413)
FZR is a 7WD domain family member implicated in cell cycle regulation of Drosophila and yeast. It has been shown that the over expression of fzr proteins increases natural killer cell-mediated cell death and as a result, suppresses tumor growth. The fzr protein is essential for cyclosome activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC20 homolog 1; CDC20-like 1b; CDC20-like protein 1; Cdh1/Hct1 homolog; cell division cycle 20 related 1; Fizzy-related protein homolog; Fzr; hCDH1
Gene Aliases: AW108046; CDC20C; CDH1; FYR; FZR; FZR1; FZR2; HCDH; HCDH1; KIAA1242
UniProt ID: (Human) Q9UM11, (Mouse) Q9R1K5
Entrez Gene ID: (Human) 51343, (Mouse) 56371, (Rat) 314642
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support