Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive Control: K562 cell lysate, Siha cell lysate, rat liver tissue, human liver tissue, human placenta tissue, mouse liver tissue.
Subcellular Location: Secreted.
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 µg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Christmas factor; coagulation factor 9; Coagulation factor IX; Coagulation factor IX (plasma thromboplastic component, Christmas disease, hemophilia B); EC 3.4.21.22; factor 9; factor IX F9; Factor9; FIX F9; plasma thromboplastic component; Plasma thromboplastin component; PTC; RP6-88D7.1; serine protease
Gene Aliases: AW111646; Cf-9; Cf9; F9; F9 p22; FIX; HEMB; P19; PTC; THPH8
UniProt ID: (Rat) P16296, (Human) P00740, (Mouse) P16294
Entrez Gene ID: (Rat) 24946, (Human) 2158, (Mouse) 14071
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support