Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: GKDELFALEQ SCAQVVLQAA NERNVSTRQG MDLSANQDEE TDQETFQLEI DRDTKKCAFR THTGKYWTLT ATGGVQSTAS SKNASCYFDI EWRDRRITLR ASNGKFVTSK KNGQ
Fascin organizes filamentous actin into bundles with a minimum of 4. 1:1 actin/fascin ratio. It is probably involved in the assembly of actin filament bundles present in microspikes, membrane ruffles, and stress fibers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 55 kDa actin-bundling protein; 55kDa actin-bundling protein; Fascin; fascin 1; Fascin homolog 1; fascin homolog 1, actin bundling protein; fascin homolog 1, actin-bundling protein; Fascin1; fscn1 {ECO:0000250|UniProtKB:Q16658}; p55; Singed; Singed (Drosophila) like (sea urchin fascin homolog like); singed-like (fascin homolog, sea urchin); Singed-like protein
Gene Aliases: AI663989; FAN1; Fascin; fascin-1; FSCN1; HSN; p55; SNL
UniProt ID: (Human) Q16658, (Rat) P85845, (Mouse) Q61553
Entrez Gene ID: (Human) 6624, (Rat) 683788, (Mouse) 14086
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support