Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The Rhesus Fc Receptor Binding Inhibitor can be used to inhibit the non-specific Fc receptor (FcR)-mediated binding of mouse monoclonal antibodies used for flow cytometric analysis of rhesus tissue. FcR are expressed at varying levels in multiple cell lineages including high expression in myeloid cells, granulocytes, and B cells. The biological function of the FcR, including initiation of endocytosis, phagocytosis, and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to rhesus FcR varies depending on the isotype of the monoclonal antibody.
eBioscience testing indicates that mouse IgG2a isotype is blocked by Rhesus Fc Receptor Binding Inhibitor.
Applications Reported: Rhesus Fc Receptor Binding Inhibitor has been reported for use in flow cytometric analysis.
To inhibit the non-specific binding of mouse monoclonal antibodies add the Rhesus Fc Receptor Binding Inhibitor to samples and incubate on ice for 20 minutes. Without washing, proceed to stain with primary antibody.
Applications Tested: Rhesus Fc Receptor Binding Inhibitor has been pre-titrated and tested by flow cytometric analysis of normal rhesus peripheral blood cells. This can be used at 5 µL (10 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
Four different classes of Fcy receptors have been identified: FcyRI (CD64), FcyRII (CD32), and FcyRIII (CD16), and they are expressed at varying levels in multiple cell lineages including high expression in myeloid, granulocyte and B cells. The biological function of the FcyR, including initiation of endocytosis, phagocytosis and antigen presentation, is elicited upon binding of host-immunoglobulin. The extent to which mouse monoclonal antibodies will bind to human FcyR varies depending on the isotype of the monoclonal antibody. Furthermore, different monoclonal antibodies of the same isotype will display different binding to human FcyRs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fc blocking; FcR BI; FcR Inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support