Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 16G2; fetuin beta; fetuin phosphoprotein; Fetuin-B; Fetuin-like protein IRL685; Gugu; IRL685
Gene Aliases: 16G2; 2310011O17Rik; AI255764; D17980; Fet; FETUB; Gugu; IRL685; Pp63
UniProt ID: (Human) Q9UGM5, (Mouse) Q9QXC1, (Rat) Q9QX79
Entrez Gene ID: (Human) 26998, (Mouse) 59083, (Rat) 83928
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support