Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Positive Samples: Human plasma, Rat liver
Immunogen sequence: NDEGEGEFWL GNDYLHLLTQ RGSVLRVELE DWAGNEAYAE YHFRVGSEAE GYALQVSSYE GTAGDALIEG SVEEGAEYTS HNNMQFSTFD RDADQWEENC A
Fibrinogen is a soluble protein found in blood plasma and is produced by the liver cells. The protein is a hexamer consisting of two sets of disulfide linked chains. Fibrinogen is essential for blood coagulation as the precursor of fibrin. It is converted to fibrin by the action of thrombin in the presence of calcium ions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fibrinogen a chain; Fibrinogen alpha; Fibrinogen alpha chain; Fibrinogen α chain; fibrinogen, A alpha polypeptide; fibrinogen, alpha polypeptide; Fibrinogenα chain
Gene Aliases: Ac1873; ENSMUSG00000059807; Fba5e; FGA; Fib; Fib2
UniProt ID: (Human) P02671, (Mouse) E9PV24
Entrez Gene ID: (Human) 2243, (Mouse) 14161, (Rat) 361969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support