Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Immunogen sequence: HDDQDYLHSL GKARTAQVQK DARIGEAEAK RDAGIREAKA KQEKVSAQYL SEIEMAKAQR DYELKKAAYD IEVNTRRAQA DLAYQLQVAK TKQQIEEQRV QVQVVERAQQ VAVQEQEIAR REKELEARVR KPAEAERYKL ERLAEAEKSQ LIMQAEAEAA SVRMRGEAEA FAIGARARAE AEQMAKKAEA FQLYQEAAQL DMLLEKLPQV AEEISGPLTS ANKITLVSSG SGTMGAAKVT GEVLDILTRL PESVERLTGV SISQVNHKPL RTA
Positive Samples: HepG2, HeLa, 293T, A-549, Mouse lung, Mouse heart, Mouse brain, Rat heart, Rat brain; Cellular Location: Cell membrane, Endosome, Melanosome, Membrane, Peripheral membrane protein, caveola
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-118P11.2; Flotillin-1; integral membrane component of caveolae; OTTHUMP00000164855; REG-2; Reggie-2
Gene Aliases: FLOT1; Rareg; Reg2; reggie-2
UniProt ID: (Human) O75955, (Mouse) O08917, (Rat) Q9Z1E1
Entrez Gene ID: (Human) 10211, (Mouse) 14251, (Rat) 64665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support