Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Aliquoting is unnecessary for -20°C storage.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epidermal surface antigen; ESA; Flotillin 2 (epidermal surface antigen 1); Flotillin-2; Membrane component chromosome 17 surface marker 1; Membrane component chromosome 17 surface marker 1 homolog; membrane component, chromosome 17, surface marker 1 (35kD protein identified by monoclonal antibody ECS-1); OTTHUMP00000163513; REG-1; Reggie-1; reggie1-1
Gene Aliases: AI573412; ECS-1; ECS1; ESA; ESA1; FLOT2; M17S1; Reg1; reggie-2
UniProt ID: (Human) Q14254, (Mouse) Q60634, (Rat) Q9Z2S9
Entrez Gene ID: (Rabbit) 100344093, (Human) 2319, (Pig) 100518752, (Mouse) 14252, (Rat) 83764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support