Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA5-14710 targets Follicle Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples.
The MA5-14710 immunogen is human Follicle Stimulating Hormone.
MA5-14710 detects Follicle Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa.
Product MA514710 is a smaller package size of MIF2710 (formerly sold as a Seradyn product).
Follicle-stimulatine hormone (FSH) is a member of the glycoprotein hormone family. Members of this family consist of an identical alpha subunit and a hormone-specific beta subunit. Alternative splicing results in two transcript variants encoding the same protein. FSH is secreted by gonadotropic cells in the pituitary gland and in conjunction with luteinizing hormone, induces egg and sperm production.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: follicle stimulating hormone, beta polypeptide; Follicle-stimulating hormone beta subunit; Follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH-B; FSH-beta
Gene Aliases: FSHB; HH24
UniProt ID: (Human) P01225
Entrez Gene ID: (Human) 2488
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support